
Huperzine A

Synonym(s):
(−)-Huperzine A, (−)-Selagine, Kimpukan A
Empirical Formula (Hill Notation):
C15H18N2O
CAS Number:
Molecular Weight:
242.32
Products
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing