
Greiner CELLSTAR® multiwell culture plates

Synonym(s):
Greiner CELLSTAR® multiwell culture plates
Products
Product Category
Brand
Color
Manufacturer
Binding Type
Volume (ml)
Wells
Color
Package Size
Sterility
Showing 1-7 of 7
Compare
Product Number
Description
Pricing