
G418 disulfate salt

Synonym(s):
Antibiotic G418
Empirical Formula (Hill Notation):
C20H40N4O10 · 2H2SO4
CAS Number:
Molecular Weight:
692.71
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Product Line
Activity Spectrum
Quality Segment
Application
Mode of Action
Sterilization Method
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing