
Foscarnet sodium

Synonym(s):
Sodium phosphonoformate tribasic hexahydrate, Foscarnet, Phosphonoformic acid trisodium salt hexahydrate, Foscarnet sodium hexahydrate
Linear Formula:
Na2O3PCO2Na · 6H2O
CAS Number:
Molecular Weight:
300.04
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Package Size
API Family
Reference Material Type
Special Grade
Technique