
Ethylmagnesium bromide solution

Linear Formula:
CH3CH2MgBr
CAS Number:
Molecular Weight:
133.27
Products
Product Category
Brand
Sustainability
Feature
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Reaction Type
Showing 1-5 of 5
Compare
Product Number
Description
Pricing