
Escherichia coli WDCM 00013 Vitroids™

Synonym(s):
E. coli WDCM 00013 Vitroids™
Products
Brand
Manufacturer
Physical Form
Agency
Quality Segment
Application
Microorganism Suitability
Package Size
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing