
Disodium 5′-uridylate

Synonym(s):
Uridine 5′-monophosphate disodium salt, 5′-UMP-Na2, U 5′-P, UMP, Uridylic acid disodium salt
Empirical Formula (Hill Notation):
C9H11N2Na2O9P
CAS Number:
Molecular Weight:
368.15
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing