
Demethoxycurcumin

Synonym(s):
(E,E)-1-(4-Hydroxy-3-methoxyphenyl)-7-(4-hydroxyphenyl)-1,6-heptadiene-3,5-dione, Curcumin II, Desmethoxycurcumin, Monodemethoxycurcumin
Empirical Formula (Hill Notation):
C20H18O5
CAS Number:
Molecular Weight:
338.35
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Research Area
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3