
(+)-B-Methoxydiisopinocampheylborane

Synonym(s):
(+)-Diisopinocampheylmethoxyborane
Empirical Formula (Hill Notation):
C21H37BO
CAS Number:
Molecular Weight:
316.33
Products
Product Category
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing