
Cyanidin 3-galactoside chloride

Synonym(s):
Idaein chloride, 3-(Galactosyloxy)-3′,4′,5,7-tetrahydroxyflavylium chloride, Cyanidin 3-O-β-D-galactopyranoside, Cyanidin 3-O-galactoside chloride, Idein chloride
Empirical Formula (Hill Notation):
C21H21ClO11
CAS Number:
Molecular Weight:
484.84
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing