
CGS-21680 hydrochloride hydrate

Synonym(s):
2-p-(2-Carboxyethyl)phenethylamino-5′-N-ethylcarboxamidoadenosine hydrochloride hydrate
Empirical Formula (Hill Notation):
C23H29N7O6 · HCl · xH2O
CAS Number:
Molecular Weight:
535.98 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing