
Calmodulin from bovine testes

Synonym(s):
CaM, Phosphodiesterase 3′:5′-cyclic nucleotide activator
CAS Number:
Products
Product Category
Brand
Biological Source
Physical Form
Product Line
Purity
Tissue
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing