
Bosutinib

Synonym(s):
4-[(2,4-Dichloro-5-methoxyphenyl)amino]-6-methoxy-7-[3-(4-methyl-1-piperazinyl)propoxy]-3-quinolinecarbonitrile, SKI-606, WAY-173606
Empirical Formula (Hill Notation):
C26H29Cl2N5O3
CAS Number:
Molecular Weight:
530.45
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing