
Amrinone Related Compound A

Synonym(s):
1,6-Dihydro-6-oxo-(3,4′-bipyridine)-5-carboxamide, 5-Carboxamide[3,4′-bipyridin]-6(1H)-one
Empirical Formula (Hill Notation):
C11H9N3O2
CAS Number:
Molecular Weight:
215.21
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2