
Adapalene Related Compound D

Synonym(s):
4,4′-Dimethoxy-3,3′-di(adamant-1-yl)biphenyl
Empirical Formula (Hill Notation):
C34H42O2
CAS Number:
Molecular Weight:
482.70
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2