
3,4-Dimethyl-1H-pyrazole

Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing