
(2RS)-2-(4-methylphenyl)propanoic acid

Synonym(s):
α,4-Dimethylphenylacetic acid, 2-(4-Methylphenyl)propanoic acid, p-Methylhydratropic acid
Linear Formula:
CH3C6H4CH(CH3)CO2H
CAS Number:
Molecular Weight:
164.20
Products
Brand
Formula Weight
Melting Point (°C)
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing