Skip to Content
Merck
2-Methyl-4-isothiazolin-3-one hydrochloride ≥99%

2-Methyl-4-isothiazolin-3-one hydrochloride

2-Methyl-4-isothiazolin-3-one hydrochloride ≥99%
Synonym(s):
MIT hydrochloride, Methylisothiazolinone hydrochloride
Empirical Formula (Hill Notation):
C4H5NOS · HCl
CAS Number:
Molecular Weight:
151.61
EC Number:

Products

Brand
Formula Weight
Purity
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing