
2-Bromo-1,3,2-benzodioxaborole

Synonym(s):
B-Bromocatechol-borane
Empirical Formula (Hill Notation):
C6H4BBrO2
CAS Number:
Molecular Weight:
198.81
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing