Skip to Content
Merck
1-Amino-2-methyl-2-propanol 95% anhydrous basis

1-Amino-2-methyl-2-propanol

1-Amino-2-methyl-2-propanol 95% anhydrous basis
Synonym(s):
1-Amino-2-methylpropan-2-ol
Empirical Formula (Hill Notation):
C4H11NO
CAS Number:
Molecular Weight:
89.14

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing