Skip to Content
Merck
All Photos(1)

Key Documents

91344

Supelco

Bis(2-ethylhexyl) isophthalate

analytical standard

Synonym(s):

1,3-Benzenedicarboxylic acid 1,3-bis(2-ethylhexyl) ester, DOIP, Di-2-ethylhexyl isophthalate, Isophthalic acid bis(2-ethylhexyl ester)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H38O4
CAS Number:
Molecular Weight:
390.56
Beilstein:
2162836
UNSPSC Code:
77101502
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

InChI

1S/C24H38O4/c1-5-9-12-19(7-3)17-27-23(25)21-14-11-15-22(16-21)24(26)28-18-20(8-4)13-10-6-2/h11,14-16,19-20H,5-10,12-13,17-18H2,1-4H3

InChI key

WXZOXVVKILCOPG-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoyuki Nishikawa et al.
Molecular therapy : the journal of the American Society of Gene Therapy, 22(12), 2046-2055 (2014-07-16)
Tumor microenvironments include a number of fibrin clots due to the microbleeding caused by cancer cell invasion into blood vessels, which suggests the potential utility of a platelet vector for systemic cancer treatment. We previously reported that inactivated Sendai virus

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service