Skip to Content
Merck
All Photos(1)

Key Documents

AB3832

Sigma-Aldrich

Anti-Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558) Antibody, phospho-specific

Chemicon®, from rabbit

Synonym(s):

Anti-EBP50, Anti-NHERF, Anti-NHERF-1, Anti-NHERF1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

monkey, human, mouse, rat

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pThr567)

Gene Information

human ... SLC9A3R1(9368)

Specificity

Recognizes Thr567 phosphorylated ezrin, Thr564 phosphorylated radixin, and Thr558 phosphorylated moesin. Antibody does not cross-react with other related phospho proteins such as merlin or band 4.1.

Immunogen

Epitope: phospho-specific
KLH-conjugated, synthetic phospho-peptide corresponding to residues surrounding Thr567 of human ezrin

Application

Anti-Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558) Antibody, phospho-specific detects level of Ezrin (Thr567)/Radixin (Thr564)/Moesin (Thr558) & has been published & validated for use in IH(P) & WB.
Research Category
Cell Structure
Research Sub Category
Cytoskeleton
Western blotting 1:1,000 (for best results, incubate membrane with diluted antibody in 5% BSA, 1X TBS, 0.1% Tween-20 at 4°C with gentle shaking, overnight). The antibody recognizes 75KDa (Moesin, and ~80kDa Ezrln & Radixin) bands.

Immunohistochemistry (paraffin): 1:50

Optimal working dilutions must be determined by the end user.

Target description

75KDa Moesin, and ~80kDa Ezrin & Radixin

Physical form

ImmunoAffinity Purified
Liquid in 10 mM sodium HEPES, pH 7.5, 150 mM NaCl, 100 μg/ml BSA, with 50% glycerol.

Storage and Stability

Maintain at -20°C for 12 months, do not aliquot.

Analysis Note

Control
Cerebral cortex, basal ganglia, hippocampus, hypophysis, and optic nerve

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minji Kim et al.
Journal of cell science, 128(23), 4317-4327 (2015-10-21)
Tubulogenesis is fundamental to the development of many epithelial organs. Although lumen formation in cysts has received considerable attention, less is known about lumenogenesis in tubes. Here, we utilized tubulogenesis induced by hepatocyte growth factor (HGF) in MDCK cells, which
Fa-Min Zeng et al.
Molecular medicine reports, 14(5), 4802-4810 (2016-10-26)
The key molecular events that contribute to tumorigenesis are incompletely understood. The aim of the present study was to characterize and compare the biological phenotypes of three human telomerase reverse transcriptase (hTERT) and/or human papillomavirus 16 E6 and E7‑immortalized esophageal epithelial
Anika L Dzierlenga et al.
Journal of biochemical and molecular toxicology, 32(3), e22035-e22035 (2018-01-18)
Nonalcoholic steatohepatitis (NASH) remodels the expression and function of genes and proteins that are critical for drug disposition. This study sought to determine whether disruption of membrane protein trafficking pathways in human NASH contributes to altered localization of multidrug resistance-associated
Kyung Yong Lee et al.
Molecular cell, 68(1), 61-75 (2017-09-26)
Double-strand breaks (DSBs) of DNA in eukaryotic cells are predominantly repaired by non-homologous end joining (NHEJ). The histone chaperone anti-silencing factor 1a (ASF1a) interacts with MDC1 and is recruited to sites of DSBs to facilitate the interaction of phospho-ATM with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service