Skip to Content
Merck
All Photos(1)

Key Documents

324870

Sigma-Aldrich

Anti-α-Epithelial Cell Sodium Channel (20-42) Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-α-ENaC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human, rat

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCNN1A(6337)
rat ... Scnn1A(25122)

General description

Immunoaffinity purified rabbit polyclonal antibody. Recognizes the ~97 kDa glycosylated α-ENaC protein.
Recognizes the ~97 kDa glycosylated α-ENaC protein.
This Anti-α-Epithelial Cell Sodium Channel (20-42) Rabbit pAb is validated for use in Immunoblotting, Immunoprecipitation for the detection of α-Epithelial Cell Sodium Channel (20-42).

Immunogen

Human
a synthetic peptide [LMKGNKREEQGLGPEPAAPQQPT(C)] corresponding to amino acids 20-42 of human α-epithelial cell sodium channel (α-ENaC), conjugated to carrier protein

Application

Immunoblotting (1 µg/ml)

Immunoprecipitation (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In PBS, 1 mg/ml BSA.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Benos, D.J., and Stanton, B.A. 1999. J. Physiol. London520, 631.
Mano, I., and Driscoll, M. 1999. Bioessays21, 568.
Fyfe, G.K., et al. 1998. Semin. Nephrol.18, 138.
Does not cross-react with β-ENaC or γ-ENaC. Also reported to work for immunoprecipitation. Variables associated with assay conditions will dictate the optimal working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qian Wang et al.
The Journal of biological chemistry, 288(1), 264-273 (2012-11-20)
Epithelial sodium channel (ENaC) in the kidneys is critical for Na(+) balance, extracellular volume, and blood pressure. Altered ENaC function is associated with respiratory disorders, pseudohypoaldosteronism type 1, and Liddle syndrome. ENaC is known to interact with components of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service