Skip to Content
Merck
All Photos(1)

Key Documents

1.46777

Millipore

Phosphate Buffer Solution

bottle of, ready-to-use, bottle volume 500 mL , filling volume

Synonym(s):

Phosphate buffer, buffer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700

Agency

EP 2.6.1
JP 4.06
USP 71
71

Quality Level

sterility

sterile; autoclaved

form

liquid

feature

closure type screw cap with 3 loci
ready-to-use

packaging

bottle of

technique(s)

direct inoculation: suitable

bottle capacity

500 mL

bottle volume

500 mL , filling volume

pH

7.2 ± 0.2

application(s)

cosmetics
food and beverages
pharmaceutical

storage temp.

2-25°C

suitability

nonselective for

General description

Phosphate buffer is designed to cater as a sample diluent and a solution for performing serial dilution. It prevents pH-associated variations while carrying out microbial experiments.

Application

Phosphate buffer is used for the preparation of dilutions, blanks for the examination of waters, dairy products, foods, and other test samples.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service