Skip to Content
Merck
All Photos(1)

Key Documents

850172P

Avanti

rac-16:0PI(3,5)P2-d5

Avanti Research - A Croda Brand 850172P, powder

Synonym(s):

1,2-dipalmitoyl-rac-glycero(d5)-3-phospho-(1′-myo-inositol-3′,5′-bisphosphate) (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H85D5N3O19P3
CAS Number:
Molecular Weight:
1027.11
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 100 μg (with stopper and crimp cap (850172P-100ug))
pkg of 1 × 500 μg (with stopper and crimp cap (850172P-500ug))

manufacturer/tradename

Avanti Research - A Croda Brand 850172P

shipped in

dry ice

storage temp.

−20°C

SMILES string

[2H]C(C([2H])([2H])OP([O-])(O[C@H]1[C@H](O)[C@@H](OP(O)([O-])=O)[C@H](O)[C@@H](OP([O-])(O)=O)[C@H]1O)=O)(OC(CCCCCCCCCCCCCCC)=O)C([2H])([2H])OC(CCCCCCCCCCCCCCC)=O.[NH4+].[NH4+].[NH4+]

Application

rac-16:0PI(3,5)P2-d5 is suitable for use as an internal standard in lipid extraction for lipid mass spectrometry.

Biochem/physiol Actions

Phosphoinositol-containing sphingolipids can induce ATPase at lower levels when compared to glycerophospholipids.

Packaging

2 mL Amber Serum Vial with Stopper and Crimp Cap (850172P-100ug)
2 mL Amber Serum Vial with Stopper and Crimp Cap (850172P-500ug)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of PI4P levels by PI4KIII alpha during G-protein-coupled PLC signaling in Drosophila photoreceptors
Balakrishnan SS, et al.
Journal of Cell Science, 131(15), jcs217257-jcs217257 (2018)
Phosphatidylinositol phosphate, phosphatidylinositol bisphosphate, and the phosphoinositol sphingolipids are found in the plasma membrane and stimulate the plasma membrane H+-ATPase of Saccharomyces cerevisiae
Patton JL and Lester RL
Archives of Biochemistry and Biophysics, 292(1), 70-76 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service