Skip to Content
Merck
All Photos(1)

Key Documents

488461

Sigma-Aldrich

Carbon-13C tetrabromide

99 atom % 13C

Synonym(s):

Tetrabromomethane-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CBr4
CAS Number:
Molecular Weight:
332.62
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Assay

99% (CP)

form

solid

bp

190 °C (lit.)

mp

88-90 °C (lit.)

density

3.43 g/mL

mass shift

M+1

SMILES string

Br[13C](Br)(Br)Br

InChI

1S/CBr4/c2-1(3,4)5/i1+1

InChI key

HJUGFYREWKUQJT-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service