MilliporeSigma
HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L, Trace Level Detection
HPLC Analysis of Lactic Acid Enantiomers on Astec® CLC-L, Trace Level Detection application for HPLC
Materiales
Estándar
Referencia del producto
Descripción
Precios
D-(−)-Lactic acid
≥90%
Lactic acid
meets USP testing specifications
Ácido L-(+)-Láctico
≥98%
Ácido L-(+)-Láctico
BioXtra, ≥98% (titration)
DL-Lactic acid
~90% (T)
Ácido L-(+)-Láctico
analytical standard
Lactic acid
Pharmaceutical Secondary Standard; Certified Reference Material
columna analítica
Referencia del producto
Descripción
Precios
Astec® CLC-L Chiral HPLC Column
5 μm particle size, L × I.D. 15 cm × 4.6 mm
CONDITIONS
column
Astec CLC-L, 15 cm x 4.6 mm I.D., 5 μm particles (53123AST)
mobile phase
5 mM copper sulfate
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
10 μL
sample
5 mg/mL in water