Merck
  • On the Mechanism of Connecting Deltahedral Zintl Clusters via Conjugated Buta-1,3-dien-1,4-diyl Functionalities: Synthesis and Structure of [Ge
On the Mechanism of Connecting Deltahedral Zintl Clusters via Conjugated Buta-1,3-dien-1,4-diyl Functionalities: Synthesis and Structure of [Ge
Chemistry (Weinheim an der Bergstrasse, Germany) (2017-09-15)
Manuel M Bentlohner, Sabine Frischhut, Thomas F Fässler
RESUMEN
We report on the synthesis and structure, as well as on the mechanism of formation of the [Ge
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,4-Bis(trimethylsilyl)butadiyne, 98%, stable crystalline form of butadiyne