Merck
  • Mutation increasing β-carotene concentrations does not adversely affect concentrations of essential mineral elements in pepper fruit.
Mutation increasing β-carotene concentrations does not adversely affect concentrations of essential mineral elements in pepper fruit.
PloS one (2017-02-17)
Nasya B Tomlekova, Philip J White, Jacqueline A Thompson, Emil A Penchev, Stephan Nielen
RESUMEN
Vitamin and mineral deficiencies are prevalent in human populations throughout the world. Vitamin A deficiency affects hundreds of millions of pre-school age children in low income countries. Fruits of pepper (Capsicum annuum L.) can be a major dietary source of precursors to Vitamin A biosynthesis, such as β-carotene. Recently, pepper breeding programs have introduced the orange-fruited (of) trait of the mutant variety Oranzheva kapiya, which is associated with high fruit β-carotene concentrations, to the mutant variety Albena. In this manuscript, concentrations of β-carotene and mineral elements (magnesium, phosphorus, sulphur, potassium, zinc, calcium, manganese, iron and copper) were compared in fruit from P31, a red-fruited genotype derived from the variety Albena, and M38, a genotype developed by transferring the orange-fruited mutation (of) into Albena. It was observed that fruit from M38 plants had greater β-carotene concentration at both commercial and botanical maturity (4.9 and 52.7 mg / kg fresh weight, respectively) than fruit from P31 plants (2.3 and 30.1 mg / kg fresh weight, respectively). The mutation producing high β-carotene concentrations in pepper fruits had no detrimental effect on the concentrations of mineral elements required for human nutrition.
MATERIALES
Número de producto
Marca
Descripción del producto
2,6-Di-tert-butyl-4-methylphenol, European Pharmacopoeia (EP) Reference Standard