Merck
Constitutively active mutation of ACVR1 in oral epithelium causes submucous cleft palate in mice.
Developmental biology (2015-06-28)
Kazuo Noda, Yuji Mishina, Yoshihiro Komatsu
RESUMEN
Cleft palate is among the most common human birth defects. Submucous cleft palate (SMCP) is a subgroup of cleft palate, which may be as common as overt cleft palate. Despite the high frequency of SMCP in humans, only recently have several animal models of SMCP begun to provide insight into the mechanisms by which SMCP develops. In this study, we show that enhanced BMP signaling through constitutively active ACVR1 in palatal epithelium causes submucous cleft palate in mice. In these mutant mice, the fusion of both palatal mesenchyme in hard palate, and muscles in soft palate were hampered by epithelial tissue. During palatal fusion, enhanced SMAD-dependent BMP signaling impaired cell death and altered cell proliferation rate in medial edge epithelium (MEE), and resulted in MEE persistence. At the molecular level, downregulation of ΔNp63, which is crucial for normal palatal fusion, in MEE cells was impaired, leading to a reduction in caspase-3 activation. Our study provides a new insight into the etiology of SMCP caused by augmented BMP signaling.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-IRF6 antibody produced in rabbit, affinity isolated antibody