Merck
  • Effects of some anti-diabetic and cardioprotective agents on proliferation and apoptosis of human coronary artery endothelial cells.
Effects of some anti-diabetic and cardioprotective agents on proliferation and apoptosis of human coronary artery endothelial cells.
Cardiovascular diabetology (2012-03-23)
Linnéa Eriksson, Ozlem Erdogdu, Thomas Nyström, Qimin Zhang, Åke Sjöholm
RESUMEN
The leading cause of death for patients suffering from diabetes is macrovascular disease. Endothelial dysfunction is often observed in type 2 diabetic patients and it is considered to be an important early event in the pathogenesis of atherogenesis and cardiovascular disease. Many drugs are clinically applied to treat diabetic patients. However, little is known whether these agents directly interfere with endothelial cell proliferation and apoptosis. This study therefore aimed to investigate how anti-diabetic and cardioprotective agents affect human coronary artery endothelial cells (HCAECs). The effect of anti-diabetic and cardioprotective agents on HCAEC viability, proliferation and apoptosis was studied. Viability was assessed using Trypan blue exclusion; proliferation in 5 mM and 11 mM of glucose was analyzed using [3H]thymidine incorporation. Lipoapoptosis of the cells was investigated by determining caspase-3 activity and the subsequent DNA fragmentation after incubation with the free fatty acid palmitate, mimicking diabetic lipotoxicity. Our data show that insulin, metformin, BLX-1002, and rosuvastatin improved HCAEC viability and they could also significantly increase cell proliferation in low glucose. The proliferative effect of insulin and BLX-1002 was also evident at 11 mM of glucose. In addition, insulin, metformin, BLX-1002, pioglitazone, and candesartan significantly decreased the caspase-3 activity and the subsequent DNA fragmentation evoked by palmitate, suggesting a protective effect of the drugs against lipoapoptosis. Our results suggest that the anti-diabetic and cardioprotective agents mentioned above have direct and beneficial effects on endothelial cell viability, regeneration and apoptosis. This may add yet another valuable property to their therapeutic effect, increasing their clinical utility in type 2 diabetic patients in whom endothelial dysfunction is a prominent feature that adversely affect their survival.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Sodium palmitate, ≥98.5%