Merck
The urinary metabolomics profile of an Italian autistic children population and their unaffected siblings.
The journal of maternal-fetal & neonatal medicine : the official journal of the European Association of Perinatal Medicine, the Federation of Asia and Oceania Perinatal Societies, the International Society of Perinatal Obstetricians (2014-10-07)
Antonio Noto, Vassilios Fanos, Luigi Barberini, Dmitry Grapov, Claudia Fattuoni, Marco Zaffanello, Andrea Casanova, Gianni Fenu, Andrea De Giacomo, Maria De Angelis, Corrado Moretti, Paola Papoff, Raffaella Ditonno, Ruggiero Francavilla
RESUMEN
A supervised multivariate model to classify the metabolome alterations between autistic spectrum disorders (ASD) patients and controls, siblings of autistic patients, has been realized and used to realize a network model of the ASD patients' metabolome. In our experiment we propose a quantification of urinary metabolites with the Mass Spectroscopy technique couple to Gas Chromatography. A multivariate model has been used to extrapolate the variables of importance for a network model of interaction between metabolites. In this way we are able to propose a network-based approach to ASD description. Children with autistic disease composing our studied population showed elevated concentration of several organic acids and sugars. Interactions among diet, intestinal flora and genes may explain such findings. Among them, the 3-(3-hydroxyphenyl)-3-hydroxypropanoic acid has been previously described as altered in autistic subjects. Other metabolites increased are 3,4-dihydroxybutyric acid, glycolic acid and glycine, cis-aconitic acid; phenylalanine, tyrosine, p-hydroxyphenylacetic acid, and homovanillic acid are all involved in the tyrosine pathway leading to neurotransmitter cathecolamine. GC-MS-based metabolomic analysis of the urinary metabolome suggests to have the required sensitivity and specificity to gain insight into ASD phenotypes and aid a personalized network-based medicine approach.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
(S)-3,4-Dihydroxybutyric acid lithium salt hydrate, ≥95.0% (GC)