Merck
Comparative and functional analysis of testis-specific genes.
Biological & pharmaceutical bulletin (2011-01-08)
FuJun Liu, ShaoHua Jin, Ning Li, Xin Liu, HaiYan Wang, JianYuan Li
RESUMEN
The testis is the special male gonad responsible for spermatogenesis and steroidogenesis with complex gene expressions. Characterizing and comparing the testis-specific genes in different species can reveal key genes related to testis specific functions and provide supplementary information for study of human testis function. We screened testis-specific genes from Unigene libraries, total 317, 449 and 147 testis-specific genes were identified for human, mouse and rat, respectively. Ten from thirteen selected human testis-specific genes were validated exclusively expressed in the testis by reverse transcription polymerase chain reaction (RT-PCR). Systematic bioinformatics analysis showed that specific genes were mainly related to spermatogenesis and testis development process with significant Glycolysis and Pyruvate metabolism. Enrichment functions were discussed.