Merck
Isolation of isoflavonoids possessing antioxidant activity from the fermentation broth of Streptomyces sp.
The Journal of antibiotics (1989-09-01)
K Komiyama, S Funayama, Y Anraku, A Mita, Y Takahashi, S Omura, H Shimasaki
RESUMEN
Three antioxidant isoflavonoids characterized as 4',7,8-trihydroxyisoflavone (1), 3',4',7-trihydroxyisoflavone (2) and 8-chloro-3',4',5,7-tetrahydroxyisoflavone (3) were isolated from the cultured broth of Streptomyces sp. OH-1049. Among them, 3 is a novel isoflavonoid possessing a chlorine atom in the molecule. In in vitro studies, these antibiotics were found to possess antioxidant activity whereas showed almost no cytocidal activities against HeLa S3 cells.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
YN1, ≥98% (HPLC)