Merck
Rational engineering of Candida antarctica lipase B for selective monoacylation of diols.
Chemical communications (Cambridge, England) (2012-09-05)
Anders Hamberg, Steffen Maurer, Karl Hult
RESUMEN
The enzyme Candida antarctica lipase B was subjected to site directed mutagenesis suggested by molecular modelling. The selectivity for the enzyme increased towards a range of diols over their corresponding monoesters as an effect of the mutations.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,4-Butanediol, ReagentPlus®, 99%