- Aminothiadiazole (NSC 4728) in patients with advanced ovarian carcinoma. A phase II study of the Gynecologic Oncology Group.
Aminothiadiazole (NSC 4728) in patients with advanced ovarian carcinoma. A phase II study of the Gynecologic Oncology Group.
American journal of clinical oncology (1986-08-01)
R F Asbury, J Wilson, J A Blessing, H J Buchsbaum, P J DiSaia
PMID3751971
RESUMEN
Thirty-two evaluable patients with advanced epithelial ovarian cancer were treated with aminothiadiazole at a dosage of 125 mg/m2 weekly. Two patients had partial responses, 12 had stable disease, 16 had increasing disease, and two were inevaluable for response. Aminothiadiazole used in this dosage and schedule has minimal activity in ovarian carcinoma patients previously treated.