Merck
beta-Glucosidase in Candida albicans and its application in yeast identification.
Journal of clinical microbiology (1987-05-01)
I Polacheck, M Melamed, H Bercovier, I F Salkin
RESUMEN
In this report we attempt to explain the discrepancy between beta-glucosidase (EC 3.2.1.21) activity in Candida albicans as measured by commercial kits and that found in an experimental assay. beta-Glucosidase activity in American and Israeli isolates of C. albicans was evaluated with the API ZYM and YeastIdent systems (Analytab Products) and with experimental biochemical assays. Activity was found with whole cells and cell extracts of isolates from both sources. The greatest beta-glucosidase activity was found at pH 5.0 and with p-nitrophenyl-beta-glucopyranoside (PNP-BDG) as the substrate. In assays with beta-naphthyl-beta-D-glucopyranoside and 6-bromo-2-naphthyl-beta-D-glucopyranoside (6-Br-2-naphthyl-BDG), no enzyme activity was detected in whole cells and only limited activity was found in cell extracts of isolates from both sources. In studies with PNP-BDG at pH 5.0 and 7.5, 29 to 38% less activity was found at both pHs with American whole cells, and minor activity (20%) was found at pH 7.5 with isolates from both sources. Because assays with PNP-BDG in cell extracts of isolates from both sources showed no significant differences in activity, the more limited beta-glucosidase activity in American whole cells was most likely due to less efficient transport. Because the API ZYM system uses 6-Br-2-naphthyl-BDG as the substrate and because the substrate is buffered at pH 7.5 in the API YeastIdent kit, both systems appear to be of limited value for the detection of beta-glucosidase activity in C. albicans.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
6-Bromo-2-naphthyl β-D-glucopyranoside, ≥99% (TLC)