Merck
  • Determination of carbofuran in surface water and biological tissue by sol-gel immunoaffinity extraction and on-line preconcentration/HPLC/UV analysis.
Determination of carbofuran in surface water and biological tissue by sol-gel immunoaffinity extraction and on-line preconcentration/HPLC/UV analysis.
Talanta (2012-01-24)
Luz E Vera-Avila, Bani P Márquez-Lira, Marcos Villanueva, Rosario Covarrubias, Gustavo Zelada, Valérie Thibert
RESUMEN
A selective and simple analytical method for the trace level determination of carbofuran in complex environmental and biological samples was developed based on immunoaffinity extraction (IAE) followed by on-line preconcentration and HPLC/UV analysis of the purified extract. The immunosorbent for IAE was prepared by sol-gel encapsulation of monoclonal anti-carbofuran antibodies, and was fully characterized for capacity, repeatability, binding strength, binding kinetics and cross-reactivity. Method performance was evaluated with two different types of difficult samples: dam water and methanolic extracts of epithelial cervical-uterine tissue. Linear behavior and quantitative recoveries were obtained from the analysis of samples spiked with carbofuran at 0.2-4 ng/mL (dam water, 50 mL samples) and 10-40 ng/mL (biological tissue extract, 2 mL samples). RSD (n=7) and detection limits were, respectively, 10.1% (spike 0.40 ng/mL) and 0.13 ng/mL for dam water; 8.5% (spike 20 ng/mL) and 5 ng/mL for the biological tissue extract. The excellent sample purification achieved with the IAE column allows precise and accurate determination of carbofuran in complex matrices, even when using non-selective UV detection in the chromatographic analysis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Carbofuran, 98%
Supelco
Carbofuran, PESTANAL®, analytical standard