Merck
  • Synthesis of bis-armed amino acid derivatives via the alkylation of ethyl isocyanoacetate and the Suzuki-Miyaura cross-coupling reaction.
Synthesis of bis-armed amino acid derivatives via the alkylation of ethyl isocyanoacetate and the Suzuki-Miyaura cross-coupling reaction.
Amino acids (2006-10-13)
S Kotha, V R Shah, S Halder, R Vinodkumar, K Lahiri
RESUMEN
Two synthetic routes to bis-armed-alpha-amino acid derivatives are described. The first route involves alkylation of dibromo derivatives with ethyl isocyanoacetate under phase-transfer catalysis (PTC) conditions. The second route uses a palladium-mediated Suzuki-Miyaura cross-coupling reaction between a DL-4-boronophenylalanine derivative and aromatic diiodo (or dibromo) compounds.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cyanoacetic acid, 99%