Merck
Protein assembly line components in prodigiosin biosynthesis: characterization of PigA,G,H,I,J.
Journal of the American Chemical Society (2006-09-28)
Sylvie Garneau-Tsodikova, Pieter C Dorrestein, Neil L Kelleher, Christopher T Walsh
RESUMEN
The red streptomycete metabolite prodigiosin has a unique tripyrrolic structure with two of the three pyrrolyl moieties in tandem. Five enzymes, PigA,G,H,I, and J, are involved in dipyrrole (rings A and B) formation. We have heterologously expressed and purified from Escherichia coli these five enzymes. At first, pyrrole ring A is formed on the peptidyl carrier protein PigG by one of two possible ways: (i) by action of the adenylation domain PigI that transforms l-proline into l-prolyl-AMP and by the flavoprotein dehydrogenase PigA responsible for the four-electron oxidation reaction; (ii) by loading with the pyrrolyl-2-carboxyl-(S)-pantetheinyl moiety from synthetic pyrrolyl-CoA using the phosphopantetheinyl transferase Sfp. Subsequently, pyrrole ring B is constructed by PigH after the transfer of ring A to the ketosynthase of PigJ. PigH consists of three domains: two acyl carrier proteins (ACPs) and a seryltransferase (SerT). Using HPLC and nanospray-Fourier Transform Mass Spectrometry (nFTMS), we established that all three domains of PigH undergo post-translational modifications and gained insight into the machinery involved in 2,2-dipyrrole biosynthesis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DL-p-Hydroxyphenyllactic acid, ≥97% (HPLC)