Merck
  • Determination of paramethoxyamphetamine and other amphetamine-related designer drugs by liquid chromatography/sonic spray ionization mass spectrometry.
Determination of paramethoxyamphetamine and other amphetamine-related designer drugs by liquid chromatography/sonic spray ionization mass spectrometry.
Rapid communications in mass spectrometry : RCM (2002-04-12)
Kjell A Mortier, Riet Dams, Willy E Lambert, Els A De Letter, Serge Van Calenbergh, André P De Leenheer
RESUMEN
Paramethoxyamphetamine (PMA) is an amphetamine-like designer drug that has emerged recently on the European illicit drug market. This drug has a wicked reputation, as a number of lethal intoxications have occurred. A method using high-performance liquid chromatography coupled to ion trap based mass spectrometry (LC/MS) is described for the determination of this compound together with 3,4-methylenedioxymethamphetamine (XTC or MDMA), amphetamine and 3,4-methylenedioxyamphetamine (MDA) in human matrices. A liquid/liquid extraction (LLE) was applied to whole blood, urine and postmortem tissues. Reversed-phase liquid chromatography was performed on a narrow-bore phenyl-type column at a flow rate of 0.3 mL/min. A switch box allowed disposal of early-eluting irrelevant material to waste, protecting the mass spectrometer from contamination. The column effluent was directed into an ion trap mass spectrometer by a sonic spray ionization (SSI) interface. The method was validated for all three matrices, proving the applicability of SSI even when dealing with complex biological matrices. The within-and between-day precisions were less than 17.5% and accuracy was below 16.2%. Weighted (1/x) quadratic calibration curves were generated ranging from 10 to 1000 ng/mL (blood and urine) or 20 to 2000 ng/g (tissue) and correlation coefficients (r(2)) always exceeded 0.995. In addition, the mass spectrum of PMA is given together with a proposed fragmentation pattern for the obtained LC/MS spectrum. This information can be useful for future identification of PMA with LC/MS in biological matrices as well as in confiscated powders or tablets.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
3,4-Methylenedioxyphenethylamine hydrochloride, 98%