Merck
TRANCE, a TNF family member, is differentially expressed on T cell subsets and induces cytokine production in dendritic cells.
Journal of immunology (Baltimore, Md. : 1950) (1999-03-11)
R Josien, B R Wong, H L Li, R M Steinman, Y Choi
RESUMEN
TNF-related activation-induced cytokine (TRANCE) is a member of the TNF family recently identified in activated T cells. We report here that TRANCE mRNA is constitutively expressed in memory, but not naive, T cells and in single-positive thymocytes. Upon TCR/CD3 stimulation, TRANCE mRNA and surface protein expression are rapidly up-regulated in CD4+ and CD8+ T cells, which can be further enhanced on CD4+ T cells by CD28-mediated costimulation. However, TRANCE induction is significantly suppressed when cells are stimulated in the presence of IL-4, but is not modified in the presence of IFN-alpha, IFN-gamma, TGF-beta, TNF-alpha, or IL-2. High levels of TRANCE receptor expression are found on mature dendritic cells (DCs). In this study we show that activated T and B cells also express TRANCE receptor, but only at low levels. TRANCE, however, does not exert any significant effect on the proliferation, activation, or survival of those cells. In DCs, TRANCE induces the expression of proinflammatory cytokines (IL-6, IL-1) and T cell growth and differentiation factors (IL-12, IL-15) in addition to enhancing DC survival. Moreover, TRANCE cooperates with CD40 ligand or TNF-alpha to further increase the viability of DCs, suggesting that several TNF-related molecules on activated T cells may cooperatively regulate the function and survival of DCs to enhance T cell-mediated immune responses.