Merck
  • Protein kinase B/AKT mediates insulin-like growth factor 1-induced phosphorylation and nuclear export of histone deacetylase 5 via NADPH oxidase 4 activation in vascular smooth muscle cells.
Protein kinase B/AKT mediates insulin-like growth factor 1-induced phosphorylation and nuclear export of histone deacetylase 5 via NADPH oxidase 4 activation in vascular smooth muscle cells.
Journal of cellular physiology (2019-02-23)
Paulina Pietruczuk, Ashish Jain, Estelle R Simo-Cheyou, Madhu B Anand-Srivastava, Ashok K Srivastava
RESUMEN
Insulin-like growth factor 1 (IGF-1) mediates the generation of reactive oxygen species (ROS) and the activation of growth promoting signaling pathways. Histone deacetylases (HDACs) regulate gene transcription by deacetylating lysine residues in histone and nonhistone proteins and a heightened HDAC activation, notably of HDAC5, is associated with vascular disorders, such as atherosclerosis. Although the contribution of IGF-1 in these pathologies is well documented, its role in HDAC phosphorylation and activation remains unexplored. Here, we examined the effect of IGF-1 on HDAC5 phosphorylation in vascular smooth muscle cells (VSMCs) and identified the signaling pathways involved in controlling HDAC5 phosphorylation and nuclear export. Treatment of A10 VSMCs with IGF-1 enhanced HDAC5 phosphorylation. Blockade of the IGF-1 receptor tyrosine kinase (TK) activity with the specific pharmacological inhibitor, AG1024, significantly inhibited IGF-1-induced HDAC5 phosphorylation, whereas the epidermal growth factor receptor (EGFR) TK antagonist, AG1478, had no effect. Inhibition of the mitogen-activated protein kinase pathway with U0126, SP600125, or SB203580, did not affect HDAC5 phosphorylation, whereas two inhibitors of the phosphoinositide 3-kinase (PI3K)/AKT pathways, wortmannin and SC66, almost completely attenuated IGF-1-induced responses as confirmed by immunoblotting of phospho-HDAC5 and by small interfering RNA (siRNA)-induced AKT silencing. Moreover, the NAD(P)H oxidase (Nox) inhibitor, diphenyleneiodonium (DPI), and Nox4 siRNA, attenuated IGF-1-induced phosphorylation of HDAC5 and AKT. The HDAC5 phosphorylation resulted in its nuclear export, which was reversed by SC66 and DPI. Our results indicate that IGF-1-induced phosphorylation and nuclear export of HDAC5 involve Nox4-dependent ROS generation and PI3K/AKT signaling pathways.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-phospho-HDAC5 (pSer259) antibody produced in rabbit, affinity isolated antibody