Merck
A High-Content Screen for C/EBPα Expression Identifies Novel Therapeutic Agents in Dedifferentiated Liposarcoma.
Clinical cancer research : an official journal of the American Association for Cancer Research (2021-10-21)
Christina V Angeles, Ana Velez, Jordan Rios, Bernadette Laxa, David Shum, Penelope D Ruiz, Yawei Shen, Irina Ostrovnaya, Rodrigo Gularte-Mérida, Benjamin A Nacev, Mark A Dickson, Hakim Djaballah, Tomoyo Okada, Samuel Singer
RESUMEN
Dedifferentiated liposarcoma (DDLS), one of the most common and aggressive sarcomas, infrequently responds to chemotherapy. DDLS survival and growth depend on underexpression of C/EBPα, a tumor suppressor and transcriptional regulator controlling adipogenesis. We sought to screen and prioritize candidate drugs that increase C/EBPα expression and may therefore serve as differentiation-based therapies for DDLS. We screened known bioactive compounds for the ability to restore C/EBPα expression and inhibit proliferation selectively in two DDLS cell lines but not in normal adipose-derived stem cells (ASC). Selected hits' activity was validated, and the mechanism of the most potent, SN-38, was investigated. The in vivo efficacy of irinotecan, the prodrug of SN-38, was evaluated in DDLS xenograft models. Of 3,119 compounds, screen criteria were met by 19. Validation experiments confirmed the DDLS selectivity of deguelin, emetine, and SN-38 and showed that they induce apoptosis in DDLS cells. SN-38 had the lowest IC50 (approximately 10 nmol/L), and its pro-apoptotic effects were countered by knockdown of CEBPA but not of TP53. Irinotecan significantly inhibited tumor growth at well-tolerated doses, induced nuclear expression of C/EBPα, and inhibited HIF1α expression in DDLS patient-derived and cancer cell line xenograft models. In contrast, doxorubicin, the most common treatment for nonresectable DDLS, reduced tumor growth by 30% to 50% at a dose that caused weight loss. This high-content screen revealed potential treatments for DDLS. These include irinotecan, which induces apoptosis of DDLS cells in a C/EBPα-dependent, p53-independent manner, and should be clinically evaluated in patients with advanced DDLS.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Monoclonal Anti-Vinculin antibody produced in mouse, clone VIN-11-5, ascites fluid
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad