Merck
  • Cathepsins S, B and L with aminopeptidases display β-secretase activity associated with the pathogenesis of Alzheimer's disease.
Cathepsins S, B and L with aminopeptidases display β-secretase activity associated with the pathogenesis of Alzheimer's disease.
Biological chemistry (2011-05-19)
Israel Schechter, Etty Ziv
RESUMEN
β-site APP-cleaving enzyme (BACE1) cleaves the wild type (WT) β-site very slowly (k(cat)/K(m): 46.6 m(-1) s(-1)). Therefore we searched for additional β-secretases and identified three cathepsins that split the WT β-site much faster. Human cathepsin S cleaves the WT β-site (k(cat)/K(m): 54 700 m(-1) s(-1)) 1170-fold faster than BACE1 and cathepsins B and L are 440- and 74-fold faster than BACE1, respectively. These cathepsins split two bonds flanking the WT β-site (K-MD-A), where the K-M bond (85%) is cleaved more efficiently than the D-A bond (15%). Cleavage at the major K-M bond yields Aβ (amyloid β-peptide) extended by N-terminal Met that should be removed to generate Aβ initiated by Asp1. The activity of cytosol and microsomal aminopeptidases on relevant peptides revealed rapid removal of N-terminal Met but not N-terminal Asp. Brain aminopeptidases showed similar specificity. Thus, aminopeptidases would convert Aβ extended by Met into regular Aβ (Asp1) found in amyloid plaques. Earlier studies indicate that Aβ is likely produced in the endosome and lysosome system where cathepsins S, B and L are localized and cysteine cathepsin inhibitors reduce the level of Aβ in cells and animals. Taken together, cathepsins S, B and L deserve further evaluation as therapeutic targets to develop disease modifying drugs to treat Alzheimer's disease.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Cathepsin S Active human, recombinant, expressed in FreeStyle 293-F cells, ≥90% (SDS-PAGE)