Merck
PTEN Is Required for The Anti-Epileptic Effects of AMPA Receptor Antagonists in Chronic Epileptic Rats.
International journal of molecular sciences (2020-08-13)
Ji-Eun Kim, Hana Park, Ji-Eun Lee, Tae-Hyun Kim, Tae-Cheon Kang
RESUMEN
α-Amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid receptor (AMPAR) is one of the ligand-gated ion channels for glutamate, which is an important player in the generation and spread of seizures. The efficacy of AMPAR functionality is regulated by the trafficking, synaptic targeting, and phosphorylation. Paradoxically, AMPAR expression and its phosphorylation level are decreased in the epileptic hippocampus. Therefore, the roles of AMPAR in seizure onset and neuronal hyperexcitability in ictogenesis remain to be elucidated. In the present study, we found that AMPAR antagonists (perampanel and GYKI 52466) decreased glutamate ionotropic receptor AMPA type subunit 1 (GRIA1) surface expression in the epileptic rat hippocampus. They also upregulated phosphatase and tensin homolog deleted on chromosome 10 (PTEN) expression and restored to basal levels the upregulated phosphoinositide 3-kinase (PI3K)/AKT1 phosphorylations. Dipotassium bisperoxovanadium(pic) dihydrate (BpV(pic), a PTEN inhibitor) co-treatment abolished the anti-epileptic effects of perampanel and GYKI 52466. Therefore, our findings suggest that PTEN may be required for the anti-epileptic effects of AMPAR antagonists.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-β-actina monoclonal antibody produced in mouse, clone AC-74, ascites fluid
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
Anti-GluR2 Antibody, from rabbit, purified by affinity chromatography
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad