Merck
  • Split luciferase-based assay for simultaneous analyses of the ligand concentration- and time-dependent recruitment of β-arrestin2.
Split luciferase-based assay for simultaneous analyses of the ligand concentration- and time-dependent recruitment of β-arrestin2.
Analytical biochemistry (2019-03-12)
Timo Littmann, Armin Buschauer, Günther Bernhardt
RESUMEN
Functional selectivity of agonists has gained increasing interest in G protein-coupled receptor (GPCR) research, e.g. due to expectations of drugs with reduced adverse effects. Different agonist-dependent GPCR conformations are conceived to selectively activate a balanced or imbalanced intracellular signalling response, involving e.g. different Gα subtypes, Gβγ-subunits and β-arrestins. To discriminate between the different signalling pathways (bias), sensitive techniques are needed that do not interfere with signalling. We applied split luciferase complementation to the GPCR/β-arrestin2 interaction and thoroughly analysed the influence of its implementation on intracellular signalling. This led to an assay enabling the functional characterization of ligands at the hH1R, the hM1,5R and the hNTS1R in live HEK293T cells. As demonstrated at the hM1,5R, the assay was sensitive enough to identify iperoxo as a superagonist. Time-dependent analyses of the recruitment of β-arrestin2 became possible, allowing the identification of class A and class B GPCRs, due to the differential duration of their interaction with β-arrestin2 and their recycling to the cell membrane. The developed β-arrestin2 recruitment assay, which provides concentration- and time-dependent information on the interaction between GPCRs and β-arrestin2 upon stimulation of the receptor, should be broadly applicable and of high value for the analysis of agonist bias.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Gqα- and G₁₁α-Subunits, Internal (283-300) Rabbit pAb, liquid, Calbiochem®