- Protection against anthrax toxemia by hexa-D-arginine in vitro and in vivo.
Protection against anthrax toxemia by hexa-D-arginine in vitro and in vivo.
Infection and immunity (2003-12-23)
Miroslav S Sarac, Juan R Peinado, Stephen H Leppla, Iris Lindberg
PMID14688144
RESUMEN
The anthrax toxin protective antigen precursor is activated by proteolytic cleavage by furin or a furin-like protease. We present here data demonstrating that the small stable furin inhibitor hexa-D-arginine amide delays anthrax toxin-induced toxemia both in cells and in live animals, suggesting that furin inhibition may represent a reasonable avenue for therapeutic intervention in anthrax.
MATERIALES