Merck
  • Correction: Regulation of E-cadherin localization by microtubule targeting agents: rapid promotion of cortical E-cadherin through p130CAS/Src inhibition by eribulin.
Correction: Regulation of E-cadherin localization by microtubule targeting agents: rapid promotion of cortical E-cadherin through p130CAS/Src inhibition by eribulin.
Oncotarget (2019-08-07)
Nicholas F Dybdal-Hargreaves, April L Risinger, Susan L Mooberry
RESUMEN
[This corrects the article DOI: 10.18632/oncotarget.23798.].
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anticuerpo anti-E-caderina, clon DECMA-1, clone Decma-1, from rat