Merck
  • SPINK3 modulates mouse sperm physiology through the reduction of nitric oxide level independently of its trypsin inhibitory activity.
SPINK3 modulates mouse sperm physiology through the reduction of nitric oxide level independently of its trypsin inhibitory activity.
Reproduction (Cambridge, England) (2012-01-10)
L Zalazar, T E Saez Lancellotti, M Clementi, C Lombardo, L Lamattina, R De Castro, M W Fornés, A Cesari
RESUMEN
Serine protease inhibitor Kazal-type (SPINK3)/P12/PSTI-II is a small secretory protein from mouse seminal vesicle which contains a KAZAL domain and shows calcium (Ca(2+))-transport inhibitory (caltrin) activity. This molecule was obtained as a recombinant protein and its effect on capacitated sperm cells was examined. SPINK3 inhibited trypsin activity in vitro while the fusion protein GST-SPINK3 had no effect on this enzyme activity. The inactive GST-SPINK3 significantly reduced the percentage of spermatozoa positively stained for nitric oxide (NO) with the specific probe DAF-FM DA and NO concentration measured by Griess method in capacitated mouse sperm; the same effect was observed when sperm were capacitated under low Ca(2+) concentration, using either intracellular (BAPTA-AM) or extracellular Ca(2+) (EDTA) chelators. The percentage of sperm showing spontaneous and progesterone-induced acrosomal reaction was significantly lower in the presence of GST-SPINK3 compared to untreated capacitated spermatozoa. Interestingly, this decrease was overcome by the exogenous addition of the NO donors, sodium nitroprusside (SNP), and S-nitrosoglutathione (GSNO). Phosphorylation of sperm proteins in tyrosine residues was partially affected by GST-SPINK3, however, only GSNO was able to reverse this effect. Sperm progressive motility was not significantly diminished by GST-SPINK3 or BAPTA-AM but enhanced by the addition of SNP. This is the first report that demonstrates that SPINK3 modulates sperm physiology through a downstream reduction of endogenous NO concentration and independently of SPINK3 trypsin inhibitory activity.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
IgG anti-conejo (molécula completa)-FITC antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-SPINK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution